Learn More
Cytiva HiTrap™ ANX Sepharose Fast Flow IEX Columnns
Weak anion exchanger for small scale purification of higher molecular weight protein
Brand: Cytiva 17-5162-01
1488.65 SEK valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic system
- High flowrates and good scale-up potential
Specifications
HiTrap ANX FF (High Sub) | |
<4 mL/min. | |
4°C to 30°C in 20% Ethanol | |
1 mL |
0.2 in. | |
HiTrap ANX (high sub) FF is prepacked with ANX (high sub) Sepharose Fast Flow, and is a weak anion exchanger for small scale purification of higher molecular weight protein as well as screening of binding and elutions conditions. | |
7 mm | |
1 x 5 mL |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.