Learn More
Thermo Scientific™ Shandon™ Holder dispense hopper Thermo Scientific Shandon
Brand: Thermo Scientific™ Shandon™ A78210002
Additional Details : Weight : 1.00000kg
Description
Included with unit: CD-ROM with unlicenced demo copy of Thermo Scientific Shandon MicroWriter™ software; Dispenser hoppers – six included; Pre-filled cassette tube sample pack; MicroWriter™ specific RS232 cable; Power cord; Operator manual.Uses foil tape and a heated stylus to imprint characters that maintain legibility even through processing solutions
- Made to withstand the workload of busy laboratories, marking approximately 20,000 characters per roll of tape
- Printing can be inverted for reading in either direction
- Change cassette colour and style easily with fully automated dispensing and labelling process
- Easy to change hoppers provide the flexibility of allowing an unlimited number of colours and cassettes
- Imprints, letters, numbers, Roman numerals and special characters on the cassettes in four font sizes
- Ability to print barcodes on cassettes
- Prints on all 45° flat-faced cassettes
- Choice of collection systems makes organising cassettes a seamless operation in any laboratory environment