missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
1422.00 SEK - 4180.00 SEK
Specifications
Antigen | HPR |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17713304
|
Invitrogen
PA5121903 |
20 μL |
1422.00 SEK
20µL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17703304
|
Invitrogen
PA5121902 |
100 μL |
4180.00 SEK
100µL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a haptoglobin-related protein that binds hemoglobin as efficiently as haptoglobin. Unlike haptoglobin, plasma concentration of this protein is unaffected in patients with sickle cell anemia and extensive intravascular hemolysis, suggesting a difference in binding between haptoglobin-hemoglobin and haptoglobin-related protein-hemoglobin complexes to CD163, the hemoglobin scavenger receptor. This protein may also be a clinically important predictor of recurrence of breast cancer.Specifications
HPR | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P00739 | |
3250 | |
Asn70-Val332 (Accession P00739), with N-terminal His Tag | |
Primary | |
-20°C | |
HPR |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
A-259H10.2; Haptoglobin-related locus; haptoglobin-related protein; HP; HPR | |
HPR | |
IgG | |
Antigen affinity chromatography | |
Antibody |