man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Heaters and Heating Mantles
Heating Tapes Cords and Mats
Horst™ HS Model Heating Cable
10789442
Print
Horst™ HS Model Heating Cable
Diameter: 159mm
Brand:
Horst™ 36004
View more versions of this product
Product Code.
10789442
5904.20 SEK
/ Each
Quantity
Estimated Shipment: 22-10-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Certificates
Accessories and Spare Parts
Specifications
Dimensions (L x W)
500mm
Model
HMSG
Product Type
Heating Mat
Dimensions (L x W x H)
500mm
Length (Metric) Power Cord
1 m