Learn More
HSPB1 Monoclonal Antibody (OTI8C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813404S
This item is not returnable.
View return policy
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
HSPB1 Monoclonal antibody specifically detects HSPB1 in Human, Non-human Primate samples. It is validated for Western BlotSpecifications
HSPB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P04792 | |
Mouse | |
Protein A/G | |
RUO | |
3315 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI8C7 | |
Unconjugated | |
HSPB1 | |
27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25 | |
Full length human recombinant protein of human HSPB1 (NP_001531) produced in E.coli. | |
30 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.