missing translation for 'onlineSavingsMsg'
Learn More

HSPB1 Monoclonal Antibody (OTI8C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody

2671.10 SEK - 5969.90 SEK

Specifications

Antigen HSPB1
Clone OTI8C7
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17528264
Encompass_Preferred
View Documents
Origene Technologies
TA813404S
30 μL
2671.10 SEK
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17508264
Encompass_Preferred
View Documents
Origene Technologies
TA813404
100 μL
5969.90 SEK
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

HSPB1 Monoclonal antibody specifically detects HSPB1 in Human, Non-human Primate samples. It is validated for Western Blot
Specifications

Specifications

HSPB1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25
Full length human recombinant protein of human HSPB1 (NP_001531) produced in E.coli.
Primary
-20° C, Avoid Freeze/Thaw Cycles
HSPB1
OTI8C7
Western Blot
Unconjugated
Mouse
Human, Non-human Primate
P04792
3315
IgG1
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.