missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human AP-2 gamma Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF5059
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AP-2 gamma Polyclonal specifically detects AP-2 gamma in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| AP-2 gamma | |
| Polyclonal | |
| Unconjugated | |
| Q92754 | |
| TFAP2C | |
| E. coli-derived recombinant human AP-2 gamma Leu128-Val223 Accession # Q92754 | |
| 100 μg | |
| Primary | |
| Detects human AP-2 gamma in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) AP-2 alpha is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Activating enhancer-binding protein 2 gamma, AP2 gamma, AP2-gamma, ERF1, estrogen receptor factor 1, hAP-2g, TFAP2C, TFAP2G, transcription factor AP-2 gamma, transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma), transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma), Transcription factor ERF-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering