missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human CCR5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1802R-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifikationer
| CCR5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CCR5 in direct Flow cytometry. | |
| Human | |
| Lyophilized |
| Flow Cytometry, CyTOF | |
| CTC5R | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| CHO Chinese hamster ovary cell line transfected with human CCR5 | |
| 25 μg | |
| 1234.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering