Learn More
Ansell Edmont™ HyFlex™ 11-628 Series Gray Polyurethane Medium Weight Gloves
Product Code. 12620752
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy
Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces. The Lycra™ in the liners of the gloves as well as in the polyurethane coating provides excellent flexibility and fit.
  • Industrial gloves
  • Special formulation of the polyurethane coating gives the gloves flexibility and fit, as well as secure grip on dry to slightly oily pieces for sure handling in wide range of environments
  • Unique combination of Dyneema™, Nylon, Lycra™, and glass fiber
  • Ultra strong fibers provide extreme resistance to cuts and burrs
  • Delivers perfect balance of comfort and sure handling with superior cut resistance
  • For safe use in controlling, handling and assembly applications involving contacts with sharp pieces
  • Lycra™ in the liners of these gloves, as well as the polyurethane coating, provides excellent flexibility and fit
Applications:
  • Assembling and mounting components
  • Assembly & inspection of components
  • Cutting glasses, sheets, panels
  • Handling materials with sharp or rough edges
  • Handling parts with sharp, rough edges
  • Handling rough machined metal parts
  • Picking, fastening components
  • Press injection molding
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 4342
Cuff Material Polyurethane
Product Type Medium Weight Gloves
Size 8
Glove Cuff Style Long Knitwrist
Liner Material Dyneema™
Lining Lycra™/Nylon/Dyneema™
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To High cut resistance