Learn More
Tocris Bioscience™ Ibrutinib
Potent and selective BTK inhibitor (IC50= 0.5 nM)
Brand: Tocris Bioscience™ 6813/50
6400.70 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Ibrutinib is a potent and selective BTK inhibitor (IC50= 0.5 nM). Selective for BTK against a screening panel of kinase enzymes. Inhibits autophosphorylation of BTK, phosphorylation of PLCγ, and phosphorylation of ERK (IC50 of 11 nM, 29 nM and 13 nM, respectively). Also inhibits ErbB4 (IC50 = 0.25-3.4 nM). Induces cytotoxicity of chronic lymphocytic leukemia (CLL) cells. Reduces clinical arthritis scores in a mouse model.Specifications
1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one | |
Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase | |
C25H24N6O2 | |
98% |
936563-96-1 | |
50 mg | |
440.5 |