Läs mer
IgA Secretory Chain Antibody (Mc29-12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64756H
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgA Secretory Chain Monoclonal antibody specifically detects IgA Secretory Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifikationer
| IgA Secretory Chain | |
| Monoclonal | |
| HRP | |
| P01876 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Mc29-12 | |
| PBS | |
| DKFZp686C15213, DKFZp686K04218, DKFZp686L19235, DKFZp686M08189, FLJ41981, FLJ43089, FLJ44381, IGH, IGH.1alpha, IGHDY1, immunglobulin heavy chain variable region, immunoglobulin heavy locus, MGC70893, MGC72071, MGC88774 | |
| Made to Human IgA | |
| 0.1 mL | |
| Immunology | |
| 3492 | |
| Store at 4°C in the dark. | |
| IgG1 |