Läs mer
Eurofins DiscoverX IGF-1R Protein, Activated
Beskrivning
Insulin-like growth factor (IGF) binding to the IGF receptors initiates an intracellular signaling cascade that leads to changes in gene expression and cell biology. Although multiple receptors for the IGFs have been identified, the IGFs primarily exert their biologic effects through ligation of the IGF1R (type I IGF receptor tyrosine kinase, JTK13). IGF1R exerts mitogenic, cell survival, and insulin-like activities, is involved in postnatal growth physiology, and is connected to proliferative disorders such as breast cancer.
GenBank X04434
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5 mM β-glycerophosphate, 1 mM Na3VO4. Frozen solution. |
| Molekylvikt (g/mol) | 48 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Renhet eller kvalitetsklass | ≥73% |
| Protein | IGF-1R, activated |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.