Learn More
Isoquinoline-4-carbaldehyde, 97%, Thermo Scientific™
595.00 SEK - 1635.00 SEK
Chemical Identifiers
CAS | 22960-16-3 |
---|---|
Molecular Formula | C10H7NO |
Molecular Weight (g/mol) | 157.172 |
MDL Number | MFCD00829440 |
InChI Key | RNQQJLYJLDQGGL-UHFFFAOYSA-N |
Synonym | isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci |
PubChem CID | 10868870 |
IUPAC Name | isoquinoline-4-carbaldehyde |
SMILES | C1=CC=C2C(=C1)C=NC=C2C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10451684
|
Thermo Scientific Maybridge
CC04904CB |
250 mg | Amber Glass Bottle |
595.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10706905
|
Thermo Scientific Maybridge
CC04904DA |
1 g | Amber Glass Bottle |
1635.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
22960-16-3 | |
157.172 | |
RNQQJLYJLDQGGL-UHFFFAOYSA-N | |
10868870 | |
C1=CC=C2C(=C1)C=NC=C2C=O |
C10H7NO | |
MFCD00829440 | |
isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci | |
isoquinoline-4-carbaldehyde |
Specifications
22960-16-3 | |
100.0 | |
C10H7NO | |
isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci | |
C1=CC=C2C(=C1)C=NC=C2C=O | |
157.172 | |
157.17 | |
Isoquinoline-4-carbaldehyde |
96.5 | |
97% | |
MFCD00829440 | |
RNQQJLYJLDQGGL-UHFFFAOYSA-N | |
isoquinoline-4-carbaldehyde | |
10868870 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.