Läs mer
Ki-67 Mouse, anti-Human, R718, Clone: B56, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566963
Beskrivning
The B56 monoclonal antibody specifically binds to the Ki-67 antigen that is expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). During the G0 phase, the antigen cannot be detected. During interphase of the cell cycle, it is associated with nucleolar components, and it is on the surface of the chromosomes during M phase. Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear. Because of the strict association of Ki-67 expression with cell proliferation, anti-Ki-67 antibodies are useful for the identification, quantification, and monitoring of growing cell populations.
Specifikationer
| Ki-67 | |
| Monoclonal | |
| R718 | |
| 100 Tests | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| B56 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.