Läs mer
Kinase buffer V (5X), Thermo Scientific™
Kvantitet:
100 ml
250 ml
Förpackningsstorlek:
100ml
250ml
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4
Specifikationer
Specifikationer
| Kemiskt namn eller material | Kinase buffer V (5X) |
| Fysisk form | Liquid |
| Kvantitet | 250 ml |
| Formulering | 100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4 |
For Research Use Only