missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Lactic Acid, 88–90%, ACS Reagent Grade, Scharlab

Product Code. 11358464
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in the food industry.

  • Synonyms: 2-Hydroxypropanoic acid, Lactol
  • Molecular Formula: C3H6O3
  • Molar Mass: 90.08g/mol
  • CAS: 79-33-4
  • EIN ECS No.: 200-018-0
  • Ignition Temperature: 380°C
  • Insoluble in C2H5OC2H5: Passes test
  • Aldehydes: Passes test

Specifications

CAS 79-33-4
Melting Point 18°C
Boiling Point 122°C (20hPa)
Molecular Formula C3H6O3
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
Solubility Information Miscible in water at 20°C
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILES CC(C(=O)O)O
IUPAC Name (2S)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 90.078
PubChem CID 107689
ChEBI CHEBI:422
Formula Weight 90.08g/mol
Percent Purity 88% min (acidimetric)
Grade ACS Reagent
Density 1.21g/cm3 
Quantity 1 L
Flash Point 112°C
Residue after Ignition 0.01% max
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Lactic acid, 88 - 90%, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-(+)-lactic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.