missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ LanthaScreen™ Eu-anti-GFP Antibody, 2.5 μg

These Europium-labeled anti-GFP antibodies bind specifically to GFP-tagged proteins allowing precise interrogation of protein samples.

Brand:  Invitrogen™ A14173

Produktkod. 12393543

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

These Europium-labeled anti-GFP antibodies bind specifically to GFP-tagged proteins allowing precise interrogation of protein samples. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.

LanthaScreen™ Eu-anti-GFP Antibody Lets You:

  • Indirectly label proteins tagged with GFP
  • Measure the right kinase
  • Interrogate active and non-activated kinases

Eu-anti-GFP Gives You the Benefits of Indirect Labeling

LanthaScreen™ Europium-labeled anti-GFP antibodies can be used for indirect labeling of proteins tagged with GFP. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein negatively impacts protein activity.

Use Eu-anti-GFP Antibody in the Highly Specific LanthaScreen™ Eu Kinase Binding Assay

When LanthaScreen™ Eu-anti-GFP Antibody is used in the LanthaScreen™ Eu Kinase Binding Assay, any signal is the result of a specific interaction of the Eu-anti-GFP antibody with the kinase of interest. Other kinases or phosphatases present without tag or with a different tag produce no signal. Unlike activity assays, the kinase binding assay can be performed with either active or non-active kinase preparations, making it ideal for detection of Type II inhibitors.

TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

GFP
Europium
2.5 μg
Target and Lead Identification and Validation
1 Each
Liquid
GFP
Kinase Assay
LanthaScreen™ Eu-anti-GFP Antibody, 2.5μg, GFP Antigen, Europium Conjugate, TR-FRET Technique, HEPES Buffered Saline Containing 0.1% Triton X-100 Storage Buffer, Used for Pharma and Biopharma, Target and Lead Identification and Validation
RUO
Primary
Europium labeled anti-GFP antibody in HEPES buffered saline containing 0.1% Triton X-100.

Store at -20°C.
Dry Ice
LanthaScreen
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.

For Research Use Only. Not for use in diagnostic procedures.