Läs mer
Invitrogen™ LGI1 Monoclonal Antibody (N283/7), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545678
Beskrivning
Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ~50% identity with LGI2, LGI3 and LGI4. 1 μg/mL of MA5-45678 was sufficient for detection of LGI1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S283-7.
The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as a candidate tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Two isoforms of LGI1 are known to exist.
Specifikationer
| LGI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| O95970, Q8K4Y5, Q9JIA1 | |
| LGI1 | |
| Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N283/7 | |
| FITC | |
| LGI1 | |
| ADLTE; ADPAEF; ADPEAF; BB130740; EPITEMPIN; Epitempin 1; Epitempin-1; EPT; ETL1; IB1099; leucine rich glioma inactivated 1; leucine-rich glioma inactivated 1; leucine-rich glioma-inactivated protein 1; leucine-rich repeat LGI family, member 1; leucine-rich, glioma inactivated 1; LGI1; OTTHUMP00000020121; OTTHUMP00000020122; UNQ775/PRO1569 | |
| Mouse | |
| Protein G | |
| RUO | |
| 252892, 56839, 9211 | |
| 4°C, store in dark | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.