Learn More
Litmus Solution, Pure, Indicator Grade, Fisher Chemical™
Product Code. 10284340
Click to view available options
Quantity:
500 mL
2.5 L
Litmus Solution, Pure, Indicator Grade
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 131528
Specifications
Packaging Amber glass bottle
Quantity 500 mL
Physical Form Liquid
Product Identifier
  • Litmus