missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Longifolene, MP Biomedicals™
Click to view available options
Quantity:
1 mL
5 mL
Specifications
Specifications
Color | Colorless |
Density | 0.928 g/mL at 25°C |
Quantity | 5 mL |
Formula Weight | 204.36 |
Physical Form | Liquid |
Chemical Name or Material | (+)-Longifolene |
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'