Läs mer
3M™ Partikelrespirator, 8300-serien
Handla allt 3M Produkter
Filtertyp:
FFP1
FFP2
FFP3
Med utandningsventil:
Ja
No
Yes
Förpackningsstorlek:
Paket med 10
Paket med 80
Beskrivning
Provide comfortable, effective respiratory protection for use in industries where workers will be exposed to dust particles and/or non-volatile liquid particles.
- Tested and certified to AS/NZS 1716:2003
- Robust outer shell improves collapse resistance
- M-Noseclip adjusts easily for fewer pressure points and greater comfort on the nose
- Braided headbands improve comfort on the neck, face, and head with a secure feel
- 3M™ Advanced Electret Filter Material gives effective filtration with low breathing resistance for consistent high-quality performance
- The super-soft, cushioned inner lining helps workers stay comfortable
- Models that include the Cool Flow™ exhalation valve offer improved comfort in hot humid environments and/or where work is hard and physical
- Colored headbands for easy identification: yellow for P1, blue for P2
Specifikationer
Specifikationer
| Certifikat och efterlevnad | AS/NZS 1716:2003; European Standard EN149:2001 + A1:2009 NR |
| Filtertyp | FFP2 |
| Material | Thermoplastic Elastomer |
| Produkttyp | General Purpose Particulate Respirator |
| Med utandningsventil | Yes |
| För användning med (applikation) | Industries where workers will be exposed to dust particles or non-volatile liquid particles |
| Latexfri | Yes |
| Modell | 8322 |
| Nosstycke typ | Aluminum clip |
| Sterilitet | Non-sterile |
| Visa mer |