Läs mer
Brand™ Melamine Formaldehyde Mortar with Pestle
Produktkod. p-8010207 Handla allt Brand Produkter
Diameter (metrisk) Yttre:
125 mm
150 mm
Höjd (metrisk):
80 mm
90 mm
Förpackningsstorlek:
Varje
Produktkod. 10495031
Brand: Brand™ 110705
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Resistant to boiling and autoclavable (at 121°C).
  • White in color
  • With foot and spout
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifikationer
Autoklaveringsbar Autoclavable
Diameter (metrisk) Yttre 125 mm
Produkttyp Mortar and Pestle
Material Melamine Formaldehyde
Höjd (metrisk) 80 mm