missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Menhaden Oil Diet, High, Powder

Menhaden oil diet. Contains AIN-76, casein, menhaden oil and mineral and vitamin mix.

6360.00 SEK - 24014.30 SEK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11412941
View Documents
MP Biomedicals
0296019410
10 kg
6360.00 SEK
10kg
Please to purchase this item. Need a web account? Register with us today!
11422941
View Documents
MP Biomedicals
0296019420
20 kg
11052.62 SEK
20kg
Please to purchase this item. Need a web account? Register with us today!
11432941
View Documents
MP Biomedicals
0296019450
50 kg
24014.30 SEK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Special procedures are required for the proper preparation of menhaden oil diets since these polyunsaturated oils rapidly oxidize upon exposure to air. Under normal conditions, an unacceptable level of oxidation occurs after just three days of exposure to the atmosphere. In order to minimize oxidation, MP Biomedicals packages the diets under nitrogen in sealed bags. It is recommended that these diets be kept frozen and accordingly, the products are shipped on dry ice.
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Powder
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.