missing translation for 'onlineSavingsMsg'
Learn More

Methyl potassium malonate, 98%, Thermo Scientific Chemicals

Product Code. 15419169
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Acts as an inhibitor in protein glycosylation and advanced glycosylation

Methyl potassium malonate is a protein advanced glycosylation inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl potassium malonate is a protein advanced glycosylation inhibitor.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38330-80-2
Molecular Formula C4H5KO4
Molecular Weight (g/mol) 156.178
MDL Number MFCD00014021
InChI Key WWTULTKUWBKVGV-UHFFFAOYSA-M
Synonym potassium 3-methoxy-3-oxopropanoate, methyl potassium malonate, potassium methyl malonate, propanedioic acid, monomethyl ester, potassium salt, potassium monomethyl malonate, monomethyl malonate potassium salt, hydrogen methyl malonate potassium salt, monomethylpotassiummalonate, acmc-209vp4, ksc226a9d
PubChem CID 2724687
IUPAC Name potassium;3-methoxy-3-oxopropanoate
SMILES COC(=O)CC(=O)[O-].[K+]

Specifications

Melting Point ∽190°C (decomposition)
Color White
Quantity 500 g
Solubility Information Soluble in methanol.
Formula Weight 156.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl potassium malonate, Hygroscopic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'