Learn More
Ansell Edmont™ Microgard™ 3000 Protective Oversleeves
6235.23 SEK valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Description
- Multi-layer barrier laminate
- Elasticated at both ends
- Reaches to mid-bicep
Specifications
Size | Universal |
Product Type | Chemical-resistant Sleeve |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Seam Type | Ultrasonically Welded |
Cuff Style | Elastic |
Certifications/Compliance | Category I PPE according to European Directive 89/686/EEC |