missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
VEGF Mouse anti-Human, Clone: 26503, R&D Systems™

Mouse Monoclonal Antibody has been used in 53 publications
Brand: R&D Systems MAB293-500
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
VEGF Monoclonal specifically detects VEGF in Human, Primate samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifikationer
| VEGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7422 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization, ELISA | |
| 26503 | |
| Western Blot 1 ug/mL, Neutralization 10-60 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_001165097.1 | |
| VEGFA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 Ala27-Arg191 Accession # NP_001165097.1 | |
| 500 μg | |
| Primary | |
| Detects human VEGF 165 and human VEGF 121 in ELISAs and Western blots. In ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) VEGF and recombinant rat (rr)VEGF and no cross-reactivity with recombinant human (rh) VEGF-D is obseved. | |
| Human, Primate | |
| Supernatant |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering