missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA
Mouse IL-13 Culture Media Quantikine ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ SM1300CB
Additional Details : Weight : 2.91000kg
Description
The Mouse IL-13 Culture Media Quantikine ELISA quantifies mouse IL-13 in cell culture supernates (50 ul).Specifications
7.80 - 500pg/mL | |
HRP | |
Cell Culture Supernatant | |
Sufficient materials to run ELISAs on at least six 96-well plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1.5pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Natural and Recombinant Mouse IL-13 | |
Mouse | |
16163 | |
Cell Culture Supernates (50uL) | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600 |