missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Mouse TSLP

Produktkod. 15567536
Klicka för att se tillgängliga alternativ
Kvantitet:
25 μg
5 μg
Förpackningsstorlek:
25µg
5µg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15567536

Brand: Invitrogen™ 14849862

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate. Description: Thymic Stromal Lymphopoietin (TSLP) is a member of the B cell-stimulating factor family. Originally identified from the conditioned medium of a mouse thymic stromal cell line that promoted B-cell development, TSLP is expressed in thymus, spleen, kidney, lung, and bone marrow. TSLP activity overlaps with that of IL-7, playing a key role in the development of B cells, as well as stimulating the growth of thymocytes and T cells. TSLP stimulates monocytes to produce T cell-attracting chemokines such as TARC (CCL17) and MDC (CCL22). TSLP also activates dendritic cell-primed CD4 T cells to produce Th2 cytokines, including IL-4, IL-5, IL-13, and TNFalpha. This hematopoietic cytokine binds to a heterodimeric receptor complex consisting of the IL-7R alpha chain (IL-7Ralpha) and the TSLP-specific chain (TSLPR). Binding induces activation of STAT3 and STAT5 via phosphorylation. Mouse TSLP shares approximately 43% amino acid sequence identity with human TSLP. Recombinant mouse TSLP is a 15.0 kDa protein consisting of 121 amino acid residues. Applications Reported: Recombinant mouse TSLP is biologically active. Applications Tested: This recombinant mouse TSLP has been tested in a Nag8/7 cell proliferation assay with an observed ED50 of 0.15ng/mL, corresponding to a specific activity of approximately 6x10e6 Units/mg. Source: Insect cell expressed N-terminal His-Tagged amino acids Tyr20-Glu140 of mTSLP accession &Num;NM_021367. Bioactivity: The ED50 tested in a Nag8/7 cell proliferation assay was 0.15ng/mL, corresponding to a specific activity of approximately 6x10e6 Units/mg. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >99% as determined by SDS-PAGE. Molecular Weight: 15-22 kDa on SDS-PAGE due to glycosylation. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer Q9JIE6
Koncentration 0.1mg/mL
För användning med (applikation) Control, Functional Assay
Formulering PBS with 1% BSA and no preservative; pH 7.2
Gen-ID (Entrez) 53603
Molekylvikt (g/mol) 15-22kDa on SDS-PAGE due to glycosylation.
Namn Mouse TSLP
pH-intervall 7.2
Reningsmetod SDS-PAGE
Kvantitet 5 μg
Källa Insect cells
Förvaringskrav -80°C
Regulatorisk status RUO
Strukturell form Insect cell expressed N-terminal His-Tagged amino acids Tyr20-Glu140 of mTSLP accession #NM_021367
Endotoxinkoncentration Less than 0.1ng/μg cytokine as determined by the LAL assay.
Vanligt namn Mouse TSLP
Gensymbol Tslp
Biologisk aktivitet The ED50 tested in a Nag8/7 cell proliferation assay was 0.15ng/mL, corresponding to a specific activity of approximately 6x106 Units/mg.
Produkttyp Recombinant Protein
Specifik reaktivitet Mouse
Konjugera Unconjugated
Korsreaktivitet Ms
Art Baculovirus
Rekombinant Recombinant
Form Liquid
Protein subtyp Recombinant
Renhet eller kvalitetsklass Affinity chromatography
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.