missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Albumin, Bovine, For IVD, Low IgG

Prepared by heat shock fractionation

1913.30 SEK - 11638.80 SEK

Specifications

For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) ∽66
Name Bovine Blood Albumin Fraction V
pH Range 7
Source Bovine Blood
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494206
View Documents
MP Biomedicals™
08810034
500g
11638.80 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
11474206
View Documents
MP Biomedicals™
08810032
50g
1913.30 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
11484206
View Documents
MP Biomedicals™
08810033
100g
3794.80 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For IVD applications, immunoassays, enzyme stabilization
  • Low protease, conductivity and visible absorbance
  • Purity: >98%
  • pH: 7

Specifications

Specifications

Immunoassay, Mammalian Cell Culture
Bovine Blood Albumin Fraction V
Bovine Blood
Crystalline
∽66
7
0° to 5°C
Videos
SDS
Documents

Documents

Product Certifications