Learn More
MP Biomedicals™ EcoChrome™ Silica Plates
Cover a wide range of Thin-Layer Chromatography applications: qualitative and quantitative separations as well as semi-preparations.Suitable for quantitative densitometric UV/VIS evaluation.
Brand:  MP Biomedicals™ 0204618
Product Code. 11303599
  • 1645.12 SEK / Pack of 2025
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EcoChrome Silica Plate
Chromatography, Chemical Synthesis, Quantitative Densitometric UV/Vis Evaluation
15°C to 30°C
20 x 20 x 0.25 cm