missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Milk Powder

Whole

Brand:  MP Biomedicals™ 0290236325

Product Code. 11495602

  • 4555.00 SEK / 25lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This is 26% butter fat whole milk in powder form. Used in formulating animal research diets.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Powder
11.33 kg
Buttmilk Fat 26%, Moisture 4%, Protein 27%, Lactose 37%, Minerals 8%, Titratable Acidity 0.15%
Used in formulating animal research diets
15°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers