Learn More
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
Brand:  MP Biomedicals™ 0215228205
 View more versions of this product
Product Code. 11418541
  • 1150.10 SEK / 5mL
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Specifications
Brown
Mouse
Mouse Serum
0.2 μm filtration
9.65 mg/dL
-20°C
Liquid
5 mL
Blocking agents and controls in immunoassays
7.4 to 8.1