Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Store, ship and package liquids with these highly reliable, lab-quality bottles, specifically designed for long-term demanding lab use.
111.04 SEK - 630.15 SEK
Specifications
Color | Translucent |
---|---|
Description | Lab Quality Narrow-Mouth Bottle |
Material | HDPE |
Sterility | Non-sterile |
Closure Color | Natural |
Product Code | Brand | Capacity (Metric) | Closure Size | Capacity (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Capacity (English) | Price | Quantity & Availability | |||
10718331
|
Thermo Scientific™
2002-9125 |
4 mL | 13 mm | 0.125 oz. |
111.04 SEK
Pack of 12 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
10550022
|
Thermo Scientific™
2002-9025 |
8 mL | 20 mm | 0.25 oz. |
121.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
15322244
|
Thermo Scientific™
2002-9025 |
8 mL | 20 mm | 0.25 oz. | |||||
10758711
|
Thermo Scientific™
2002-9050 |
15 mL | 20 mm | 0.5 oz. |
177.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10768711
|
Thermo Scientific™
2002-0001 |
30 mL | 20 mm | 1 oz. |
151.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10313501
|
Thermo Scientific™
2002-0002 |
60 mL | 20 mm | 2 oz. |
159.56 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10159140
|
Thermo Scientific™
2002-0004 |
125 mL | 24 mm | 4 oz. |
281.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10782711
|
Thermo Scientific™
2002-0006 |
175 mL | 24 mm | 6 oz. |
270.20 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10507341
|
Thermo Scientific™
2002-0008 |
250 mL | 24 mm | 8 oz. |
312.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10749671
|
Thermo Scientific™
2002-9016 |
500 mL | 38-430 mm | 16 oz. |
630.15 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10517341
|
Thermo Scientific™
2002-0016 |
500 mL | 28 mm | 16 oz. |
399.39 SEK
Pack of 12 |
In Stock
Log in to see stock. |
|||
10027950
|
Thermo Scientific™
2002-0032 |
1 L | 38-430 mm | 32 oz. |
325.43 SEK
Pack of 6 |
In Stock
Log in to see stock. |
|||
Description
Store, ship, and package liquids using highly reliable and durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
- Versatile and indispensable in the lab
- Uniform walls provide durability and resistance to splitting or punctures
- Can be used in a freezer up to -100°C (-149°F)
- Linerless polypropylene closures ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
- Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
- Leakproof† when used with Nalgene closures
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
Translucent | |
HDPE | |
Natural | |
Yes | |
Round | |
Polypropylene | |
No |
Lab Quality Narrow-Mouth Bottle | |
Non-sterile | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Narrow | |
Unlined | |
Screw Cap |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.