Learn More
Naphthalene-2-thiocarboxamide, 97%, Thermo Scientific™
Product Code. 11471200
Click to view available options
Quantity:
250 mg
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6967-89-1
Molecular Formula C11H9NS
Molecular Weight (g/mol) 187.26
MDL Number MFCD01571356
InChI Key ZQGJZFKITDDUEH-UHFFFAOYSA-N
Synonym 2-naphthalenecarbothioamide, maybridge1_008735, acmc-20ak77, 2-naphthalene carbothioamide, naphthalene-2-thiocarboxamide, amino-2-naphthylmethane-1-thione, naphthalene-2-carbothioic acid amide
PubChem CID 2800333
IUPAC Name naphthalene-2-carbothioamide
SMILES C1=CC=C2C=C(C=CC2=C1)C(=S)N
Specifications
Melting Point 150°C to 152°C
Quantity 1 g
UN Number UN2811
Formula Weight 187.26
Percent Purity 97%
Chemical Name or Material Naphthalene-2-thiocarboxamide
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'thankYou' missing translation for 'feedbackSubmitted'