missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 30% VP, Chem-Lab

Product Code. 11979232
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Used as the primary reagent for nitration and for trace analysis. Nitric Acid, 30% VP, Chem-Lab is a highly corrosive mineral acid. It is intended for laboratory use and TMAR. Appearance of a yellowish tinge in the container has no impact on product quality.

The main industrial use of nitric acid is for the production of fertilizers. Nitric acid is neutralized with ammonia to give ammonium nitrate. The other main applications are for the production of explosives, nylon precursors, and specialty organic compounds.

Specifications

pH <1
Color Colorless
CAS 7697-37-2
Concentration 386mL HNO3 65% per 1L H2O
Density 1.21g/mL
Odor Odourless
Quantity 1 L
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information Soluble in Water
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitric acid 30 weight % v.p.
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'