Learn More
Invitrogen™ Novex™ pH 3-7 IEF Buffer Kit, Includes LC5300, LC5370, LC5371
Optimized for using with Novex pH 3-7 IEF gels
Brand: Invitrogen™ LC5377
1326.73 SEK valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Includes: Each kit includes LC5300, LC5370, LC5371
Description
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Buffer Kit | |
IEF Gel | |
Novex™ |
Anode & Cathode Buffers | |
Novex IEF Gel | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.