Läs mer
Novus Biologicals™ Recombinant Human CTLA-4 His Protein
Produktkod. 18705913 Handla allt Bio Techne Produkter
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mg
Produktkod. 18705913
Brand: Novus Biologicals™ NBP222987
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 36-161 of Human CTLA-4 The Recombinant Human CTLA-4 Protein is derived from E. coli. The Recombinant Human CTLA-4 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 0.1 mg