missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Midi
Two-in-one midi kit for PCR clean-up and extraction of DNA/RNA from gels, featuring Binding Buffer NTI colored pH indicator.
Brand: Macherey-Nagel™ 740986.20
This item is not returnable.
View return policy
Description
NucleoSpin™ Gel and PCR Clean-up Midi
- Two applications in one kit – one buffer with optimal performance for both applications
- High recoveries for fragments down to 50 bp
- Binding buffer with pH indicator
- Separate buffers for single stranded DNA or SDS containing samples available
- Suitable for all gel buffer systems (e.g., TAE, TBE)
- Ensures complete removal of contaminations such as: nucleoties, primers, enzymes, mineral oil, PCR additives, detergents, and dyes
- Typical downstream applications: cloning, sequencing, PCR, restriction analysis
Specifications
Room temperature (18-25 °C) | |
Silica Membrane Technology | |
Binding capacity: 75 μg | |
Centrifuge | |
NucleoSpin Gel and PCR Clean-up Midi Columns, buffers | |
A260/A280: 1.75 - 1.85 | |
25 min/6 preps |
Midi Spin Column | |
Agarose and Polyacrylamide Gels, PCR Reaction Mixture | |
Elution volume: 200 - 400 μL | |
PCR clean-up and gel extraction | |
20 Preps | |
< 4 mL PCR mixture; < 4 g gel | |
Typical recovery: 70 - 95 % |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- NucleoSpin Gel and PCR Clean-up Midi(20)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanate