Läs mer
Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing
Kulturområde:
25,280 cm2
2528 cm²
6320 cm²
Lager av fack:
4
10
40
Förpackningsstorlek:
Paket med 10
Paket med 2
Paket med 6
Beskrivning
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells using Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing. Flow system helps ensure a controlled, contamination-free atmosphere in each culture tray by equally distributing the user-specified gas mixture. Available in 4-, 10- and 40-tray models.
- Achieve greater yields with controlled cell culture environments equally distributed within each layer
- Optimize production capacity without changing cell culture platforms
- Develop industrial scale solutions with the utilization of an active gassing Cell Factory™ Incubator
Specifikationer
Specifikationer
| Beskrivning | Cell Factory with Active Gassing |
| nr per ärende | 6 |
| Antal per förpackning | 6 |
| Produkttyp | System with Active Gassing |
| Sterilitet | Sterile |
| Kulturområde | 6320 cm² |
| Lager av fack | 10 |
| Kvantitet | Pack/Case: 1/6 |