Learn More
Ouabain, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Selective Na+, K+-ATPase inhibitor. Cardiotonic agent. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.08 μM).
Specifications
CAS | 630-60-4 |
Molecular Formula | C29H44O12 |
Quantity | 100 mg |
Synonym | ouabain, g-strophanthin, ouabaine, astrobain, gratibain, purostrophan, strodival, oubain, ouabagenin l-rhamnoside, ouabain anhydrous |
InChI Key | LPMXVESGRSUGHW-HBYQJFLCSA-N |
SMILES | CC1C(C(C(C(O1)OC2CC(C3(C4C(CCC3(C2)O)C5(CCC(C5(CC4O)C)C6=CC(=O)OC6)O)CO)O)O)O)O |
IUPAC Name | 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one |
Molecular Weight (g/mol) | 584.659 |
PubChem CID | 439501 |
ChEBI | CHEBI:472805 |