missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Pantothenic Acid Deficient Diet, Modified, Pellet

Pantothenic acid deficient diet for rabbits. Lacks pantothenic acid.

Brand:  MP Biomedicals 0296015920

 View more versions of this product

Product Code. 11482851

This item is not returnable. View return policy

Description

Description

  • Pantothenic acid control diet for rabbits
  • Lacks pantothenic acid
  • Contains casein, methionine, mineral mix and MP Bio vitamin fortification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
20 kg
Room Temperature
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.