missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17407-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL2 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. PDCL2 has been shown to interact with the chaperonin CCT and is essential for cytoskeletal protein function and cell cycle progression.Specifications
PDCL2 | |
Polyclonal | |
Unconjugated | |
PDCL2 | |
GCPHLP, PDCL2, phosducin like 2, Phosducin like protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132954, 498352, 79455 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78Y63, Q8N4E4 | |
Pdcl2 | |
PDCL2 Fusion Protein Ag11427 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction