Learn More
Heathrow Scientific Pearl™ Adjustable Volume Pipette
The Pearl™ is a single-channel pipette offering adjustable volume and precision liquid handling.
Brand:  Heathrow Scientific HS120100
2147.00 SEK valid until 2024-12-31
Use promo code "21924" to get your promotional price.
 View more versions of this product
Product Code. 16320696
  • 2260.00 SEK / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Description
The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl™ is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digital volume display
  • Labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified
  • Autoclavable
Specifications
Specifications
CE
Pearl™ Adjustable Volume Pipette
White
Reusable
HS120100
Pearl™
Adjustable Volume Pipette
3-Position, Digit
1 Box
7 cm
2.54 cm
26.04 cm
White
Autoclavable
Low-Force
ABS
1
Non-sterile
5 to 50 μL
Hook
2.75 in.
1.00 in.
10.25 in.