missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pectolyase Y-23 (A. Niger)
Highly purified enzyme from Aspergillus niger used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues.
Brand: MP Biomedicals™ 08320971
View more versions of this product
This item is not returnable.
View return policy
Description
- Contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity
- Included is a maceration stimulating factor which stimulates tissue maceration by both pectinases
- One unit is defined as an increase in A235 by 1.0 in the reaction mixture per minute
Specifications
>1000 units per gram (pectin lyase) | |
Powder | |
1 g |
Store at 2 to 8°C | |
Isolation of biologically active protoplasts |