Learn More
MP Biomedicals™ Pectolyase Y-23 (A. Niger)
Highly purified enzyme from Aspergillus niger used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues.
8795.00 SEK - 44930.00 SEK
Specifications
Purity | >1000 units per gram (pectin lyase) |
---|---|
Content And Storage | Store at 2 to 8°C |
Format | Powder |
For Use With (Application) | Isolation of biologically active protoplasts |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16349845
|
MP Biomedicals™
08320971 |
1 g |
8795.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16359845
|
MP Biomedicals™
08320972 |
10 g |
44930.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity
- Included is a maceration stimulating factor which stimulates tissue maceration by both pectinases
- One unit is defined as an increase in A235 by 1.0 in the reaction mixture per minute
Specifications
>1000 units per gram (pectin lyase) | |
Store at 2 to 8°C | |
Powder | |
Isolation of biologically active protoplasts |