missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
PELP1 Antibody (JE02-42), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32734
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PELP1 Monoclonal antibody specifically detects PELP1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifikationer
| PELP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1-2ug/sample, Immunohistochemistry-Paraffin 1:1000 | |
| HMX3, MNARmodulator of nongenomic activity of estrogen receptor, Modulator of non-genomic activity of estrogen receptor, P160, proline and glutamic acid rich nuclear protein, proline, glutamate and leucine rich protein 1, proline, glutamic acid and leucine rich protein 1, proline-, glutamic acid- and leucine-rich protein 1, proline-, glutamic acid-, leucine-rich protein 1, Transcription factor HMX3 | |
| Synthetic peptide. (Uniprot: Q8IZL8) | |
| 100 μL | |
| Cellular Markers, Transcription Factors and Regulators | |
| 27043 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| JE02-42 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering