missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol-d6, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Phenol-d6 | 13127-88-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 13127-88-3, 67-56-1 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.1, 99.9 |
Quantity | 1 mL |
CAS Max % | 0.1, 99.9 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Molecular Formula | C6H6O |
MDL Number | MFCD00002144 |
InChI Key | ISWSIDIOOBJBQZ-QNKSCLMFSA-N |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Phenol-d6
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'