Läs mer
Invitrogen™ Phospho-IKK epsilon (Ser172) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564585
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Phospho-IKK epsilon (Ser172) Polyclonal Antibody detects endogenous levels of IKK epsilon only when phosphorylated at Ser172.
IKBKE is an activator of nuclear factor-kappa-B. Over expression of IKBKE has been observed in breast cancer cells, and suppression of IKBKE induces cell death. NF-kappa-B is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa-B mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kappa-B is associated with Ikappa-B proteins in the cell cytoplasm, which inhibit NF-kappa-B activity. Ikappa-B is phosphorylated by Ikappa-B kinase (IKK) complex that contains IKK-a, IKK-b, and IKK-gamma. A novel molecule in the IKK complex was recently identified and designated IKK-iota/IKK-i. IKK epsilon is required for the activation of NF-kappa-B by PMA and T cell receptors but not by TNF-a and IL-1. IKK-iota/IKK-i message is expressed in a variety of tissues and is inducible by TNF-a, IL-1, and LPS.
Specifikationer
| Phospho-IKK epsilon (Ser172) | |
| Polyclonal | |
| Unconjugated | |
| IKBKE | |
| AW558201; I-kappa-B kinase epsilon; Ikbke; IKKE; IKK-E; IKKepsilon; IKK-epsilon; IKKI; IKK-i; IKK-related kinase epsilon; inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56489, 9641 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14164, Q9R0T8 | |
| IKBKE | |
| A synthesized peptide derived from human IKK epsilon (Phospho-Ser172). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |