missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p38 MAPK (Thr180, Tyr182) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44684G
This item is not returnable.
View return policy
Description
This antibody reacts with human and rat (100% homologous) p38 alpha, beta, and gamma. HEK 293 cells +/- UV irradiation treatment and PC12 cells +/- Sorbitol were used as positive controls. Purified from rabbit serum by sequential epitope-specific chromatography, this product contains enough material for 10 mini-blots. The antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated p38. The final product is generated by affinity chromatography using a p38-derived peptide that is phosphorylated at threonine 180 and tyrosine 182.
Phospho-p38 MAPK (Thr180, Tyr182) Polyclonal antibody specifically detects Phospho-p38 MAPK (Thr180, Tyr182) in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-p38 MAPK (Thr180, Tyr182) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P70618, Q16539 | |
MAPK14 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human p38 that contains threonine 180 and tyrosine 182. This region is conserved among many species including mouse, rat, dog, monkey and carp. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MAPK14 | |
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; Csbp2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; Dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; Exip; fk28c03; hm:zeh1243; Hog; I79_016605; LD24658p; MAP 14; MAP kinase 1; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 11; MAPK 14; MAPK 14A; MAPK1 antibody; Mapk11; Mapk14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1432, 81649 | |
-20°C | |
Liquid |