Learn More
Cole-Parmer™ Polypropylene Manifold
Expand your flow and line capacity
Brand:  Cole-Parmer™ 31522-17
 View more versions of this product
Product Code. 11715258
  • 1080.00 SEK / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Constructed of durable polypropylene.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
Polypropylene manifold; 6 outlets; 1/2 in. inlet NPT(F)
17.15 cm
0°C
3.81 cm
150 psi
99°C
Polypropylene
32°F
1/2 in.
3.81 cm
210°F